Learn More
Kallikrein 2 Mouse anti-Human, Clone: 426723, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB4104-SP
Description
Kallikrein 2 Monoclonal specifically detects Kallikrein 2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
| Kallikrein 2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3.4 ug/mL | |
| P20151 | |
| KLK2 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 2 Ile25-Pro261 Accession # P20151 | |
| 25 μg | |
| Primary | |
| Detects human Kallikrein 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12, 13, 14, 15, or B1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Neutralization | |
| 426723 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, Klk1c2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2, Ton | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3817 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.