Learn More
Robin™ Kahn Complete Shaker
Click to view available options
Speed Range:
100 rpm
180 rpm
40 to 120 rpm
Unit Size:
Each
Description
- Mixes or agitates ingredients and solutions in a variety of containers including, but not limited to, the following:
- Flasks
- Bottles
- Beakers
- Kahn racks
Specifications
Specifications
| Load Bearing Capacity (Metric) | 6.5 kg |
| Speed Range | 40 to 120 rpm |
| Electrical Requirements | 230 V, 50 Hz, 25 W |
| Wattage | 25 W |
| Frequency | 50 Hz |
| Depth (Metric) | 250 mm |
| Dimensions (D x W x H) Interior | 250 x 360 x 130 mm |
| Height (Metric) | 130 mm |
| Width (Metric) | 360 mm |
| Model | Kline Stirrer Complete 40-120T |