Learn More
Kartell™ K70 Silicon Hoses
Product Code. 10023501
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
2 mm
3 mm
4 mm
5 mm
6 mm
7 mm
8 mm
9 mm
Unit Size:
10m
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Unit Size
10023501 6 mm 10m
10726352 2 mm 10m
15712855 3 mm 10m
10736352 4 mm 10m
15722855 5 mm 10m
10207351 8 mm 10m
15742855 9 mm 10m
15732855 7 mm 10m
10535371 1 mm 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10023501 Supplier Kartell™ Supplier No. 0392500
  • ONLINE EXCLUSIVE PRICE
    44.00€
    39.46€ / 10m
    Save 4.54€
    10% Off
 Restricted Item
In Stock
Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.
  • Flexible and transparent
  • FDA, USP, Class VI, A 50p60, EPC VI.1.3.2 approved
  • Can withstand temperatures of -60°C to 200°C
Specifications
Certifications/Compliance FDA, USP, Class VI, A 50p60 and EPC VI.1.3.2 Approved
Diameter (Metric) Inner 6 mm
Material Silicone
Diameter (Metric) Outer 9 mm
Overall Length (Metric) 10 m
Min. Operating Temperature (Metric) -60°C
Packaging Quantity 10 m
Color Transparent
Max. Operating Temperature (Metric) 200°C