Learn More
Justrite™ Steel Spill Tray
Product Code. 13497152
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. 13497152
Brand: Justrite™ 10177
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Supports up to a 5 gal. (19L) safety can with 1 quart spill capacity
  • Low profile, large diameter surface to contain spills and overfill from safety cans
  • Constructed of 24 gauge coated steel
Specifications
Product Type Spill Tray
Description Coated Steel
For Use With (Equipment) Up to 5 gal. safety can with 1 qt. spill capacity
Material Steel
Height (English) 1.25 in.
Height (Metric) 3.17 cm
Weight (English) 4 lb.
Weight (Metric) 1.8 Kg