Learn More
Justrite™ Rolling Cart for Safety Cabinets
Click to view available options
For Use With (Equipment):
Safety Cabinets
Unit Size:
Each
Description
- Fits any cabinet footprint of 43W x 18 in.D (109 x 45.7cm) with a maximum height of 44 in. (111.8cm), such as Justrite 30 gal. (114L) or piggyback cabinets
- Two swivel and two fixed casters, each with brakes
- Built-in grounding wire can be attached to cabinet
- Ergonomic, removable push/pull handle
- Black powder paint for durability
Specifications
Specifications
| For Use With (Application) | To move materials to point-of-use |
| For Use With (Equipment) | Safety Cabinets |
| Height (English) | 44 in. |
| Height (Metric) | 111.76 cm |
| Includes | 2 Swivel and 2 fixed casters each with brakes, removable push or pull handle |
| Product Type | Rolling Cart |