Learn More
Justrite™ Parts Basket for Steel Cleaning Tank
Product Code. 11908039
Click to view available options
Capacity (English):
2 gal.
Unit Size:
Each
Product Code. 11908039
Brand: Justrite™ 27901Z
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Facilitates quick washing of small parts
  • Round, steel basket
  • For use with 3.79L (1 gal.) wash tank or 7.6L (2 gal.) dip tank
  • Use with Justrite Dip Tanks and Wash Tank
Specifications
Product Type Drain Basket
Capacity (English) 2 gal.
Capacity (Metric) 7.6 L
Material Steel