Learn More
Justrite™ Horizontal and Vertical Drum Vents
Click to view available options
Includes:
Operating Valve
Operating Valve, Plated Steel Flame Arrester
Operating Valve, Stainless Steel Flame Arrester
Product Type:
Horizontal Drum Vent
Vertical Drum Vent
Material:
Brass
Non-metallic Cast Iron
Unit Size:
Each
Description
- Choose brass, stainless steel or nonmetallic vent
- For petroleum-based liquids, use non-metallic or brass vents
- For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
- For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
- Has operating valve that opens automatically to prevent pressure buildup inside drum
Horizontal Drum Vent
- Has operating valve that can be opened manually to relieve internal pressure
Order Info
All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
Certifications
FM approved.
Specifications
Specifications
| Certifications/Compliance | FM |
| Product Type | Horizontal Drum Vent |
| Description | Opens at 5 psig Internal Pressure |
| For Use With (Application) | Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup |
| For Use With (Equipment) | Horizontal Drums |
| Material | Brass |
| Includes | Operating Valve |