Learn More
Justrite™ Drum Faucets, Extender, and Valve
Product Code. 11923599
Click to view available options
Product Type:
Adjustable Drum Faucet
Drum Gate Valve
Flexible Hose Extension
Rigid Drum Faucet
Material:
Brass
Stainless Steel
Unit Size:
Each
Product Code. 11923599
Brand: Justrite™ 08552
Self-closing safety drum faucets
  • Self-closing brass safety faucets feature a PTFE fluorocarbon resin seal, flame arrester and standard 0.75 in. NPT thread
  • Brass faucets screw into drum; adjustable model adjusts to spout-down position
  • Flexible faucet extensions provide accuracy, prevent spills; available in 8 in. long (20.3cm) brass and 6 in. long (15.2cm) stainless steel
  • Brass drum gate valve fits 2 in. (5.1cm) NPT openings and has wide mouth for easy dispensing of viscous liquids from drums in horizontal position
Certifications
Faucets and valve are FM approved.
Specifications
Certifications/Compliance FM
Product Type Drum Gate Valve
For Use With (Application) Easy dispensing of viscous liquids from drums in horizontal position
Material Brass