Learn More
Justrite™ Polyethylene Document Storage Tubes
Description
- Made of weather resistant polyethylene
- With twist on lid
- All boxes have designated space for a label
- Pre-marked and blank labels included in label pack
Specifications
Specifications
| Description | Tube twist on lid opening |
| Product Type | Document Storage Box |
| Color | Red |
| Material | Polyethylene |
| Includes | Label pack with 2 blank labels and labels printed SDS and instruction manual |
| For Use With (Application) | Used to keep important papers readily available while protecting them from moisture and dirt |
| Certifications/Compliance | OSHA |
| Diameter (English) Outer | 3.6 in. |
| Diameter (Metric) Outer | 9.4 cm |
| Height (English) | 12.5 in. |
| Show More |