missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Jurkat Whole Cell Lysate

Product Code. 18533993 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18533993 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18533993 Supplier Novus Biologicals™ Supplier No. NB800PC2
  • ONLINE EXCLUSIVE PRICE
    312.00€
    295.05€ / 0.5mg
    Save 16.95€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

prepared from whole tissue homogenates

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method This lysate is prepared as a positive control for separation by SDS-PAGE and subsequent Western blot analysis. Lysates are prepared in denaturing buffer WITHOUT reducing agents (i.e. 2-mercaptoethanol (BME) or dithiothreitol (DTT)). If reducing conditions are desired, add a reducing agent prior to heating. Heat lysate to 95 degrees C for 5 minutes and rapidly cool. The recommended loading amount is 0.01-0.03 mg per lane.
Quantity 0.5 mg
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Target Jurkat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.