Learn More
3M™ Jupiter™ Starter Kits
Description
- Powered unit
- Pair of Organic Vapour and Particulate (A2P) Filters and Prefilters
- Lightweight adjustable breathing tube
- 8-hour rechargeable battery (NiMH) and a battery charger
Specifications
Specifications
| Battery Life | 8 hours |
| Battery Type | NiMH |
| Filter Type | OV/HE Filter |
| Product Type | PAPR Assembly, No Headtop |
| For Use With (Application) | Protection against gases, vapors, and particulates, or a combination of contaminants per filter used |
| Includes | Powered turbo unit, 2 x A2P filters, pre-filters, breathing tube, rechargeable battery, battery charger |
| Model | RTUJUPITER |
| Quantity | 1 Ea. |