Learn More
3M™ Jupiter™ A2BEKP Filter
Product Code. 11741393
Change view
Click to view available options
:
Includes: Jupiter In Pre-Filter and Filter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11741393 Includes: Jupiter In Pre-Filter and Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11741393 Supplier 3M™ Supplier No. 7000006599
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Protects against acid gas, organic vapours, dust, mist and fumes. 3M™ Jupiter™ A2BEKP Filters are for use with the Jupiter Powered Air Respirator.
  • 2 filters per pack
  • CE approved
TRUSTED_SUSTAINABILITY
Specifications
Battery Type NiMH
Certifications/Compliance EN 12941 / CE
Design Type For Light or Heavy Duty Use
Product Type A2BEKP Filter
For Use With (Application) Protective gas, organic vapor, inorganic acids, ammoniated, particles.
For Use With (Equipment) For Jupiter A2BEKP
Quantity 12/Pk.