Learn More
Sampling Systems™ Jumbo Sack Sampler
Product Code. 15829251 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
400 mm
800 mm
Includes:
Bottle Adapter End
End Cap
Plain End
Unit Size:
Each
Product Code. 15829251
Brand: Sampling Systems™ 1309B400
Only null left
Add to basket
Add to basket
Introduced to meet the demand of sampling large items including nuts, grains and larger granules. Sampling Systems™ Jumbo Sack Sampler is a quick and efficient way to obtain samples of the substances within sacks and bags.
  • Type A: Plain style - simply hold over the bag
  • Type B: End cap - sample is held in the sampler until the sample is deposited
  • Type C: Bottle adapter - sample drops straight into the bottle
  • Manufactured from 316L stainless steel
  • Available in 400mm or 800mm lengths
Specifications
Product Type Jumbo Sampler
Material Stainless Steel (316L)
Includes End Cap
Diameter (Metric) 40 mm
Length (Metric) 400 mm