Learn More
Tocris Bioscience™ JNJ 7777120
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
JNJ 7777120 is a histamine H4 receptor antagonist; displays high affinity (Ki = 4.5 nM) and is >1000-fold selective for H4 over other histamine receptors.
Specifications
Specifications
| Chemical Name or Material | 1-[(5-Chloro-1H-indol-2-yl)carbonyl]-4-methylpiperazine |
| CAS | 459168-41-3 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C14H16ClN3O |
| Formula Weight | Observed MW: 277.75 |