Learn More
Honeywell™ Jersey Light Gloves
missing translation for 'orderingAttributeHoverText'
:
Color: Black
Description
- 5 fingers gloves in jersey fleece 100% cotton
- Straight thumb
- Knitted wrist
- The cotton interlock lining strengthens the glove's mechanical performance while absorbing hand perspiration for added comfort
- The city assembly and stretch knit provide the glove with comfort and support while ensuring dexterity and touch
- Comfortable glove providing a protection against mechanical risks and heat contact
- The brushed fabrics improves the heat insulation and comfort
- Its brown colour makes it more soilproof, thus ensuring a longer life
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY : II Intermediary risks In accordance with : STANDARDS : EN 420-03 EEC Basic standards EN 388-16 Mechanical hazards EN 407-04 Thermical hazards |
| Color | Black |
| Liner Material | Jersey Fleece Cotton |
| Length (Metric) | 26 cm |