Learn More
Cole-Parmer™ Techne Sealing Rings for Hybridization Incubators
Product Code. p-8012867
Click to view available options
For Use With (Equipment):
For Large tubes
Unit Size:
Each
Product Code. 11734466
Brand: Cole-Parmer™ 9995841
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Replacements for the Techne HB-3D Hybrigene and HB-1D Hybridiser
Specifications
For Use With (Application) For Large Tubes at less than 70°C Temperature
For Use With (Equipment) For Large tubes
Quantity 1 Each
Type Seal Ring