missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Techne Sealing Rings for Hybridization Incubators
Click to view available options
For Use With (Equipment):
For Large Tubes
Unit Size:
Each
Specifications
Specifications
| Color | Black |
| For Use With (Application) | For Large Tubes at 70°C Temperature |
| For Use With (Equipment) | For Large Tubes |
| Type | Seal Ring |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction