Learn More
Jenway™ pH Electrodes
Click to view available options
Length (Metric):
40 mm
90 mm
120 mm
150 mm
Connector Types:
2mm plug
3150/3205 waterproof connector
BNC
BNC/8 pin mini-DIN
S7
Diameter (Metric):
4.5 mm
6 mm
12 mm
Material:
Epoxy
Glass
Unit Size:
Each
Description
- Quickly reach equilibrium
- Significantly reduced response times
- Far greater reproducibility
- Significantly reduced long-term drift
Specifications
Specifications
| Connector Types | 3150/3205 waterproof connector |
| Diameter (Metric) | 12 mm |
| Type | Sealed Combination with Bulb End and Protective Cover |
| Dimensions (L x W x H) | 12 dia. x 120 mm L |
| For Use With (Application) | measurements at low temperatures or in solutions of low ionic strength, general purpose pH measurement and accuratae Q.C. or research work |
| Length (Metric) | 120 mm |
| Material | Epoxy |
| pH Range | 0 to 10 |
| Temperature (Metric) Range | 0°C to 80°C |