Learn More
Jenway™ pH Electrodes
Product Code. 13107184
Change view
Click to view available options
Length (Metric):
40 mm
90 mm
120 mm
150 mm
Connector Types:
2mm plug
3150/3205 waterproof connector
BNC
BNC/8 pin mini-DIN
S7
Diameter (Metric):
4.5 mm
6 mm
12 mm
Material:
Epoxy
Glass
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Connector Types Diameter (Metric) Material Unit Size
13107184 150 mm BNC 6 mm Glass Each
11410499 90 mm BNC 4.5 mm Glass Each
11440499 90 mm BNC 6 mm Glass Each
11440629 120 mm 2mm plug 12 mm Glass Each
13107424 120 mm 3150/3205 waterproof connector 12 mm Epoxy Each
11490619 40 mm BNC 6 mm Glass Each
11460499 120 mm BNC/8 pin mini-DIN 12 mm Epoxy Each
11449489 120 mm S7 12 mm Epoxy Each
11430629 120 mm BNC 12 mm Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 13107184 Supplier Jenway™ Supplier No. 9996041
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Connector Types BNC
Diameter (Metric) 6 mm
Type Life Science Refillable Combination with Bulb End
Dimensions (L x W x H) 6 dia. x 150 mm L
For Use With (Application) measurement of samples in test tubes and Eppendorf vessels
Length (Metric) 150 mm
Material Glass
pH Range 0 to 14
Temperature (Metric) Range 0°C to 80°C