Learn More
Jenway™ pH Electrodes
Product Code. 11460499
Click to view available options
Length (Metric):
40 mm
90 mm
120 mm
150 mm
Connector Types:
2mm plug
3150/3205 waterproof connector
BNC
BNC/8 pin mini-DIN
S7
Diameter (Metric):
4.5 mm
6 mm
12 mm
Material:
Epoxy
Glass
Unit Size:
Each
Product Code. 11460499
Brand: Jenway™ 1009075
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these electrodes for advanced applications where faster response times and greater reproducibility are required.
  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift
Specifications
Connector Types BNC/8 pin mini-DIN
Diameter (Metric) 12 mm
Type Sealed Combination with Bulb End and Protective Cover
Dimensions (L x W x H) 12 dia. x 120 mm L
For Use With (Application) standard measurements of general solutions, field measurements or in education
Length (Metric) 120 mm
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Range 0°C to 60°C
For Use With (Equipment) Jenway pH meters 3505, 3510, 3520 and 370