Learn More
Jenway™ Air Tubing
Product Code. 12778686
Change view
Click to view available options
Type:
Air tube (2 m)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12778686 Air tube (2 m) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12778686 Supplier Jenway™ Supplier No. 1008943
  • ONLINE EXCLUSIVE PRICE
    32.00€
    24.90€ / Pack of 1
    Save 7.10€
    22% Off
Estimated Shipment: 18-12-2026