Learn More
Cole-Parmer™ Jenway™ 100 Disposable Cuvettes
Product Code. 11490549
Click to view available options
Capacity (Metric):
1.5 to 3.0 mL
2.4 to 4.5 mL
Description:
UV and visible wavelengths
Visible wavelengths only
Unit Size:
Pack of 100
Product Code. 11490549
Brand: Cole-Parmer™ 9961033
Specifications
Type Cuvette
Description Visible wavelengths only
Capacity (Metric) 2.4 to 4.5 mL
Material Plastic
Quantity 100/Pk.
Path Length 10 mm
For Use With (Application) Spectral Measurement of Liquids