missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Janelia Fluor 646, Azide
GREENER_CHOICE

Product Code. 18799406
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
This item is not returnable. View return policy

Product Code. 18799406

Brand: Tocris Bioscience™ 7088/2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Red dye (Reactive group: azide). Applications: super resolution microscopy e.g. dSTORM & STED, confocal, and live cell imaging

Janelia Fluor® 646, Azide is a red fluorogenic fluorescent dye, supplied as an azide for click chemistry. Suitable for confocal fluorescent imaging and super resolution microscopy (SRM) techniques such as dSTORM (live and fixed cells) and STED imaging. Can be multiplexed for two color imaging with Janelia Fluor™ 549 SE (Cat. No. 6147). Cell permeable. Excitation maximum = 646 nm; emission maximum = 664 nm. Quantum yield = 0.54; Max. extinction coefficient = 152,000 M-1cm-1 (measured in ethanol plus 0.1% TFA); A280 correction factor is 0.19.

To measure the absorbance spectrum of this dye we recommend the following solvent: ethanol or trifluoroethanol plus 0.1% TFA.

We also offer Janelia Fluor™ conjugated antibodies and custom conjugation services with our sister company Novus Biologicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-(2-(2-(2-azidoethoxy)ethoxy)ethoxy)ethyl)carbamoyl)benzoate
Quantity 2 mg
Target Click-Reactive Janelia Fluor™ Dyes
Molecular Formula C37H44N6O6Si
Formula Weight 3535
Purity 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.