missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies JAK Inhibitor I

Product Code. 17130312
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 17130312

Brand: STEMCELL Technologies 74024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

JAK/STAT pathway inhibitor; Inhibits JAK1, JAK2, and JAK3

  • Janus Associated Kinase (JAK) Inhibitor I is a pyridine-containing tetracycle that disrupts JAK activity by interacting with the ATP binding domain
  • This inhibitor inhibits mouse JAK3 with Ki = 5 nM, JAK family members TYK2 and JAK2 with IC50 = 1 nM, and mouse JAK1 with IC50 = 15 nM, while showing weaker inhibition of other kinases (Thompson et al.)
  • Inhibits the generation of mouse induced pluripotent stem cells (Efe et al.; Kim et al.)
  • Inhibits anti-viral response associated with JAK/STAT pathway activation (Xu et al.)

Specifications

CAS 457081-03-7
Synonym CMP 6; Janus-associated kinase inhibitor I;Pyridone 6
Quantity 1 mg
Molecular Formula C18H16FN3O
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.