Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm
Product Code. 15834734
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15834734 25 mL Each
15854734 50 mL Each
15814734 100 mL Each
15844734 250 mL Each
15864734 500 mL Each
15824734 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15834734 Supplier GPE Scientific™ Supplier No. CSB/25
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Round-bottomed shape
  • Includes plain arm
  • Includes O-ring tap
Specifications
Capacity (Metric) 25 mL
Includes O-Ring Tap and Plain Arm
Product Type Vacuum/Schlenk Flask