Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket
Product Code. 15864794
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15864794 100 mL Each
15884794 25 mL Each
15804804 50 mL Each
15894794 250 mL Each
15814804 500 mL Each
15874794 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15864794 Supplier GPE Scientific™ Supplier No. JCS/100
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Estimated Shipment: 15-01-2027
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap and B14 Socket is specially designed for use with vacuum manifolds and Schlenk lines.
  • Solidly constructed for use with vacuum manifolds
  • Designed for use with Schlenk lines
  • Includes fine thread tap and B14 socket
Specifications
Capacity (Metric) 100 mL
For Use With (Equipment) Vacuum manifolds and Schlenk lines
Includes Fine thread tap and B14 socket
Product Type Sample Flask
Socket Joint Size: 14