missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ivacaftor

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
Beschreibung
Ivacaftor is a potent and selective potentiator of Cystic Fibrosis Transmembrane Conductance Regulator (CFTR) that targets F508del-CFTR and G551D-CFTR (EC50 values are 25 nM and 100 nM, respectively); its potentiation activity depends on the level of CFTR phosphorylation. Ivacaftor reversibly binds to wild type (WT-CFTR) and G551D-CFTR mutants, increasing the open probability via an ATP-independent mechanism, and enhances cAMP/PKA-signaling mediated gating activity for both WT-CFTR and G551D-CFTR mutants. In cultured human CF bronchial epithelia with G551D/F508del mutations, Ivacaftor increases Forskolin (Cat. No. 1099)-induced transepithelial current (EC50 = 236 nM) and increases chloride secretion by 10-fold. Also rescues endothelial CFTR expression and function and prevents endothelial barrier failure and protein leak in human pulmonary microvascular endothelial cells. Ivacaftor is orally bioavailable.
Spezifikation
Spezifikation
| Chemical Name or Material | N-[2,4-Bis(1,1-dimethylethyl)-5-hydroxyphenyl]-1,4-dihydro-4-oxo-3-quinolinecarboxamide |
| CAS | 873054-44-5 |
| Quantity | 50 mg |
| Target | CFTR |
| Molecular Formula | C24H28N2O3 |
| Purity | 98% |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur