missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITS Thorsten Betzel™ SenSafe™ Mercury Check Test Strip
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Description
Order Info
ForUseWithEquipment
Specifications
Specifications
| Format | Test Strip |
| For Use With (Application) | For Water Quality Testing is a Quick and Easy Method to Test for High Levels of Mercury |
| Quantity | 50 Pack |
| Test Time | 2 min. 30 sec. |
| Type | SenSafe Mercury Check Test Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction