missing translation for 'onlineSavingsMsg'
Learn More

Isoxanthopterin, TRC

Product Code. 30133287
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133287 25 mg 25mg
30004548 5 mg 5mg
30011593 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133287 Supplier Toronto Research Chemicals Supplier No. TRCI91834325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-69-1
Molecular Formula C6H5N5O2
Molecular Weight (g/mol) 179.14
IUPAC Name 2-amino-3,8-dihydropteridine-4,7-dione
SMILES NC1=NC2=C(N=CC(=O)N2)C(=O)N1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H5N5O2/c7-6-10-4-3(5(13)11-6)8-1-2(12)9-4/h1H,(H4,7,9,10,11,12,13)
Formula Weight 179.0443
Chemical Name or Material Isoxanthopterin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.