Learn More
Camping Gaz™ Isotherm Extreme 10L Cooler
Click to view available options
Capacity (Metric):
10 L
Unit Size:
Each
Description
- 30-year Guarantee
- Keeps contents cool for up to 17 hours.
- Tough outer shell
- High-quality insulation
- Integrated air-tight seal
- Lid-locking handle ensures proper seal and secures items inside.
Specifications
Specifications
| Weight (Metric) | 1.4 kg |
| Design | Traditional, with lid-locking handle |
| Type | Cooler |
| Capacity (Metric) | 10 L |