Learn More
Isoquinoline-8-carboxylic acid, 97%, Thermo Scientific Chemicals
Product Code. 10064397 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. 10064397
Brand: Thermo Scientific Acros 431050050
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61563-43-7
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD06738935
InChI Key VMNZQPRIUSJCOH-UHFFFAOYSA-N
Synonym 8-isoquinolinecarboxylic acid, 8-carboxyisoquinoline, 8-isoquinolinecarboxylicacid, 8-carboxy-2-azanaphthalene
PubChem CID 21393830
IUPAC Name isoquinoline-8-carboxylic acid
SMILES C1=CC2=C(C=NC=C2)C(=C1)C(=O)O
Specifications
Melting Point 263.0°C to 267.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 173.17
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Isoquinoline-8-carboxylic acid
compliance-icons
Product Identifier
  • Isoquinoline-8-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.