Learn More
Isopulegol, TRC
Product Code. 30407865
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407865 2.5 g 2.5g
30389815 5 g -
30393373 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30407865 Supplier Toronto Research Chemicals Supplier No. TRCI8747402.5G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7786-67-6
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
Synonym 5-Methyl-2-(1-methylethenyl)cyclohexanol, 2-Isopropenyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylvinyl)cyclohexanol, p-Menth-8-en-3-ol
IUPAC Name 5-methyl-2-prop-1-en-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(O)C1)C(=C)C
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h8-11H,1,4-6H2,2-3H3
Formula Weight 154.1358
Chemical Name or Material Isopulegol