missing translation for 'onlineSavingsMsg'
Learn More

Isoproterenol Isopropyl Ether Hydrochloride, TRC

Product Code. 30012098
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012098 25 mg 25mg
30006789 5 mg 5mg
30009685 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012098 Supplier Toronto Research Chemicals Supplier No. TRCM23139025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120285-39-4
Molecular Formula C14 H23 N O3 . H Cl
Molecular Weight (g/mol) 289.8
Synonym 4-(1-Isopropoxy-2-(isopropylamino)ethyl) Benzene-1,2-diol Hydrochloride;4-[1-(1-Methylethoxy)-2-[(1-methylethyl)amino]ethyl]-1,2-benzenediol Hydrochloride
IUPAC Name 4-[1-isopropoxy-2-(isopropylamino)ethyl]benzene-1,2-diol;hydrochloride
SMILES Cl.CC(C)NCC(OC(C)C)c1ccc(O)c(O)c1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H23NO3.ClH/c1-9(2)15-8-14(18-10(3)4)11-5-6-12(16)13(17)7-11;/h5-7,9-10,14-17H,8H2,1-4H3;1H
Formula Weight 289.1445
Chemical Name or Material Isoproterenol Isopropyl Ether Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.