Learn More
Isopropyl isothiocyanate, 97%
Product Code. 11404753
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11404753
Brand: Thermo Scientific Alfa Aesar A11641.14
Request Bulk or Custom Format
CAS: 2253-73-8 | C4H7NS | 101.167 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Isopropyl isothiocyanate is used as is an antimicrobial agent. It is also used as medicinal intermediate.
Solubility
Miscible with methanol. Hydrolyzes in water.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with alcohols and amines.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2253-73-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00037830
InChI Key VHBFEIBMZHEWSX-UHFFFAOYSA-N
Synonym isopropyl isothiocyanate, isothiocyanic acid isopropyl ester, isopropylisothiocyanate, propane, 2-isothiocyanato, unii-46561pkp9t, isopropyl mustard oil, 2-isothiocyanato-propane, propan-2-isothiocyanate, ipncs, isopropyl isothiocya
PubChem CID 75263
IUPAC Name 2-isothiocyanatopropane
SMILES CC(C)N=C=S
Specifications
Density 0.948
Boiling Point 137°C to 138°C
Flash Point 40°C (104°F)
Odor Pungent
Refractive Index 1.493
Quantity 25 g
UN Number UN2924
Beilstein 1739163
Sensitivity Moisture sensitive
Solubility Information Miscible with methanol. Hydrolyzes in water.
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Isopropyl isothiocyanate