Learn More
Isopropyl 3-oxobutanoate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 542-08-5 |
| Molecular Formula | C7H12O3 |
| MDL Number | MFCD00043933 |
| Quantity | 500 g |
| SMILES | CC(CC(OC(C)C)=O)=O |
| Molecular Weight (g/mol) | 144.17 |
| Percent Purity | 98% |
| Chemical Name or Material | Isopropyl 3-oxobutanoate |