missing translation for 'onlineSavingsMsg'
Learn More

Isophorone Diisocyanate, TRC

Product Code. 30004385
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Unit Size:
100mL
10mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004385 100 mL 100mL
30009905 50 mL 50mL
18000950 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30004385 Supplier Toronto Research Chemicals Supplier No. TRCI821978100ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-71-9
InChI Formula InChI=1S/C12H18N2O2/c1-11(2)4-10(14-9-16)5-12(3,6-11)7-13-8-15/h10H,4-7H2,1-3H3
IUPAC Name 5-isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Molecular Formula C12 H18 N2 O2
Molecular Weight (g/mol) 222.28
Chemical Name or Material Isophorone Diisocyanate

Specifications

Quantity 100 mL
Synonym Isocyanic acid, methylene(3,5,5-trimethyl-3,1-cyclohexylene) ester (7CI,8CI), 5-Isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane, 1,3,3-Trimethyl-1-(isocyanatomethyl)-5-isocyanatocyclohexane, 1-(Isocyanatomethyl)-5-isocyanato-1,3,3-trimethylcyclohexane, 1-Isocyanato-3,3,5-trimethyl-5-(isocyanatomethyl)cyclohexane, 1-Isocyanato-3-(isocyanatomethyl)-3,5,5-trimethylcyclohexane, 1-Isocyanato-5-(isocyanatomethyl)-3,3,5-trimethylcyclohexane, 3,3,5-Trimethyl-5-(isocyanatomethyl)cyclohexyl isocyanate, 3-(Isocyanatomethyl)-3,5,5-trimethylcyclohexyl isocyanate, IPDI, Isophorone diisocyanate, PDI, Z 4770, 5-Isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane, 1,3,3-Trimethyl-1-(isocyanatomethyl)-5-isocyanatocyclohexane, 1-(Isocyanatomethyl)-5-isocyanato-1,3,3-trimethylcyclohexane, 1-Isocyanato-3,3,5-trimethyl-5-(isocyanatomethyl)cyclohexane, 1-Isocyanato-3-(isocyanatomethyl)-3,5,5-trimethylcyclohexane, 1-Isocyanato-5-(isocyanatomethyl)-3,3,5-trimethylcyclohexane, 3,3,5-Trimethyl-5-(isocyanatomethyl)cyclohexyl isocyanate, 3-(Isocyanatomethyl)-3,5,5-trimethylcyclohexyl isocyanate, Carbitol acetate-isophorone Diisocyanate Copolymer, IPDI, PDI, Z 4770
SMILES CC1(C)CC(CC(C)(CN=C=O)C1)N=C=O
Formula Weight 222.1368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.