Learn More
Isonitrosoacetone, TRC
Specifications
Specifications
| CAS | 306-44-5 |
| Molecular Formula | C3H5NO2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C3H5NO2/c1-3(5)2-4-6/h2,6H,1H3/b4-2+ |
| SMILES | CC(=O)\C=N\O |
| Molecular Weight (g/mol) | 87.08 |
| Chemical Name or Material | Isonitrosoacetone |