Learn More
Isobornyl acrylate, tech. 85%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Product Code. 11322387
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322387 250 g 250g
11312387 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11322387 Supplier Thermo Scientific Alfa Aesar Supplier No. L09977.30
Request Bulk or Custom Format
It is used as UVcuring EBcuring adhesive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as UVcuring EBcuring adhesive.
Solubility
Difficult to mix in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5888-33-5
Molecular Formula C13H20O2
Molecular Weight (g/mol) 208.30
MDL Number MFCD00080424
InChI Key PSGCQDPCAWOCSH-BOURZNODSA-N
Synonym isobornyl acrylate, acrylic acid isobornyl ester, 1,7,7-trimethyltricyclo 2.2.1 hepten-2-yl-2-propenoate
PubChem CID 122130333
IUPAC Name [(3R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] prop-2-enoate
SMILES CC1(C)[C@H]2CC[C@]1(C)C(C2)OC(=O)C=C
Specifications
Melting Point <-35°C
Density 0.97
Boiling Point 104°C to 106°C (4.5 mmHg)
Flash Point 108°C (226°F)
Odor Characteristic
Refractive Index 1.4795
Quantity 250 g
UN Number UN3082
Beilstein 2254825
Sensitivity Light sensitive
Solubility Information Difficult to mix in water.
Formula Weight 208.3
Percent Purity 85%
Grade Technical
Chemical Name or Material Isobornyl acrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Isobornyl acrylate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Isobornyl acrylate