Learn More
Brady™ ISO Square Aluminum Safety Sign 'Fire Alarm Flashing Light'
Product Code. 17821785 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
100 x 100 mm
150 x 150 mm
200 x 200 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17821785 200 x 200 mm Each
17811745 100 x 100 mm Each
17821745 150 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17821785 Supplier Brady™ Supplier No. 311213
  • ONLINE EXCLUSIVE PRICE
    11.00€
    10.75€ / Pack of 1
    Save 0.25€
    2% Off
Estimated Shipment: 04-12-2026
ISO Safety Sign displaying flashing fire alarm and flames.
  • To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Alternate Part Number F/F018/NT/ALU-200X200-1
Material Aluminum
Quantity 1 Sign
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 7.87 in.
Width (English) 7.87 in.
Temperature Range (Metric) -40°C to +140°C
Shape Square
Temperature Range (English) -40°F to +284°F
Languages Universal
Dimensions (L x W) 200 x 200 mm
Displayed Text Fire Alarm Flashing Light
Product Type Safety Sign
Color White on Red