Learn More
Brady™ ISO Safety Sign without Text - Warning: Low Temperature/Freezing Conditions
Product Code. 15772652
Change view
Click to view available options
Dimensions (L x W):
43 x 50 mm
87 x 100 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15772652 43 x 50 mm Each
15871537 87 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15772652 Supplier Brady™ Supplier No. 827797
  • ONLINE EXCLUSIVE PRICE
    6.05€
    5.89€ / Pack of 1
    Save 0.16€
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Safety sign for hazard identification.
  • Sign compliant to ISO 7010
  • Improves workplace safety
Specifications
Alternate Part Number PIC W010- Tri 050-PE-SHEET/7
Finish Gloss
Material Laminated Polyester
Adhesive Type Permanent
Shape Triangular
Dimensions (L x W) 43 x 50 mm
Color Black on Yellow