Learn More
Brady™ ISO Safety Sign - Disinfect Surface, French
Product Code. 30280541
Click to view available options
Dimensions (L x W):
150 x 50 mm
148 x 210 mm
210 x 297 mm
297 x 105 mm
297 x 420 mm
450 x 150 mm
600 x 200 mm
Material:
Aluminum
Laminated Polyester
Polypropylene
Unit Size:
Each
Product Code. 30280541
Brand: Brady™ 323280
Only null left
Add to basket
Add to basket
Sign to signify that the surface must be disinfected. Text is in French.
  • Sign compliant to ISO 7010
Specifications
Certifications/Compliance ISO 7010; Compliant with Directive 2011/65/EU and 2015/863
Alternate Part Number M/M062/FR808/PP-297x420-1
Grade M062
Material Polypropylene
Quantity 1 Ea.
Mounting Type Surface Mount, Wall Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 16.54 in.
Height (Metric) 420 mm
Thickness 1.55 mm
Width (English) 11.69 in.
Width (Metric) 297 mm
Temperature Range (Metric) -40°C to +120°C
Shape Rectangular
Temperature Range (English) -40°F to +248°F
Languages French
Dimensions (L x W) 297 x 420 mm
Displayed Text Désinfectez la surface
Product Type Personal Hygiene Sign
Color White on Blue