missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isatin-5-sulfonic Acid Monosodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Isatin-5-sulfonic Acid Monosodium Salt |
| CAS | 80789-74-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C8H4NNaO5S |
| Quantity | 5 g |
| Synonym | 5-Sulfoisatin Sodium Salt, 2,3-Dihydro-2,3-dioxo-1H-indole-5-sulfonic Acid Sodium Salt, 1H-Indole-5-sulfonic acid, 2,3-dihydro-2,3-dioxo-, monosodium salt (9CI), 5-Indolinesulfonic acid, 2,3-dioxo-, sodium salt (7CI), 5-Isatinsulfonic acid sodium salt, Sodium isatin-5-sulfonate |
| InChI Formula | InChI=1S/C8H5NO5S.Na/c10-7-5-3-4(15(12,13)14)1-2-6(5)9-8(7)11;/h1-3H,(H,9,10,11)(H,12,13,14);/q;+1/p-1 |
| SMILES | O=C1C(C2=CC(S(=O)(O[Na])=O)=CC=C2N1)=O |
| IUPAC Name | sodium 2,3-dioxoindoline-5-sulfonate |
| Molecular Weight (g/mol) | 249.17 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?