missing translation for 'onlineSavingsMsg'
Learn More

Isatin-5-sulfonic Acid Monosodium Salt, TRC

Product Code. 30402775
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402775 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402775 Supplier Toronto Research Chemicals Supplier No. TRCI9006501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isatin-5-sulfonic Acid Monosodium Salt
CAS 80789-74-8
Purity Grade Notes HPLC
Molecular Formula C8H4NNaO5S
Quantity 1 g
Synonym 5-Sulfoisatin Sodium Salt, 2,3-Dihydro-2,3-dioxo-1H-indole-5-sulfonic Acid Sodium Salt, 1H-Indole-5-sulfonic acid, 2,3-dihydro-2,3-dioxo-, monosodium salt (9CI), 5-Indolinesulfonic acid, 2,3-dioxo-, sodium salt (7CI), 5-Isatinsulfonic acid sodium salt, Sodium isatin-5-sulfonate
InChI Formula InChI=1S/C8H5NO5S.Na/c10-7-5-3-4(15(12,13)14)1-2-6(5)9-8(7)11;/h1-3H,(H,9,10,11)(H,12,13,14);/q;+1/p-1
SMILES O=C1C(C2=CC(S(=O)(O[Na])=O)=CC=C2N1)=O
IUPAC Name sodium 2,3-dioxoindoline-5-sulfonate
Molecular Weight (g/mol) 249.17
Formula Weight 248.97
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.