missing translation for 'onlineSavingsMsg'
Learn More

Iron nickel oxide, tech.

Product Code. p-4407459
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11339289

Brand: Thermo Scientific Alfa Aesar 088530.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12168-54-6 | Fe2NiO4 | 234.38 g/mol

Iron nickel oxide is used as an adsorbent to remove hexavalent chromium ions present in the waste water by co-sedimentation method. It is also employed as an additive in membrane (i.e. polyvinylchloride/styrene-butadiene-rubber blend heterogeneous cation exchange membrane) fabrication, thereby increasing the permeability for bivalent ions compared to the monovalent type.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12168-54-6
Molecular Formula Fe2NiO4
Molecular Weight (g/mol) 234.38
MDL Number MFCD00016254
InChI Key NQNBVCBUOCNRFZ-UHFFFAOYSA-N
IUPAC Name oxo[(oxoferrio)oxy]iron; oxonickel
SMILES O=[Ni].O=[Fe]O[Fe]=O

Specifications

Color Brown
Physical Form Powder
Quantity 10 g
Formula Weight 234.39
Grade Technical
Odor Odorless
Appearance Dark-brown powder
Density 5.368
Chemical Name or Material Iron nickel oxide
compliance-icons compliance-icons
Product Identifier
  • Iron nickel oxide
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P363-Wash contaminated clothing before reuse.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.