Learn More
Iron(III) Perchlorate Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Iron(III) Perchlorate Hydrate |
| CAS | 15201-61-3 |
| Molecular Formula | Cl3FeH2O13 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/3ClHO4.Fe.H2O/c3*2-1(3,4)5;;/h3*(H,2,3,4,5);;1H2/q;;;+3;/p-3 |
| SMILES | O.[Fe+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O |
| IUPAC Name | iron(3+);triperchlorate;hydrate |
| Molecular Weight (g/mol) | 372.21 |
| Formula Weight | 370.791 |