Learn More
Iron(III) 2-hydroxypropane-1,2,3-tricarboxylate, BLD Pharm™
Specifications
Specifications
| CAS | 3522-50-7 |
| Molecular Formula | C6H5FeO7 |
| MDL Number | MFCD00149617 |
| Quantity | 100 g |
| SMILES | OC(C([O-])=O)(CC([O-])=O)CC([O-])=O.[Fe+3] |
| Molecular Weight (g/mol) | 244.94 |
| Chemical Name or Material | Iron(III) 2-hydroxypropane-1,2,3-tricarboxylate |