Learn More
Iron(II) Chloride Tetrahydrate, TRC
Product Code. 30396747
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396747 25 g
30394540 5 g
30393748 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30396747 Supplier Toronto Research Chemicals Supplier No. TRCI77493025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 13478-10-9
Molecular Formula FeCl2H8O4
Molecular Weight (g/mol) 198.81
Synonym Ferrous Chloride Tetrahydrate, Iron Chloride Tetrahydrate, Iron Dichloride Tetrahydrate
IUPAC Name iron(2+);dichloride;tetrahydrate
SMILES O.O.O.O.[Cl-].[Cl-].[Fe+2]
Spezifikation
Quantity 25 g
InChI Formula InChI=1S/2ClH.Fe.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2
Formula Weight 197.9149
Chemical Name or Material Iron(II) Chloride Tetrahydrate