missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) bromide, ultra dry, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11302498
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302498

Brand: Thermo Scientific Alfa Aesar 014541.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Used as a polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

Iron(II) bromide is used as a polymerization catalyst. It is used in water treatment, chemical analysis and in ultra high purity for certain crystal growth applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-46-0
Molecular Formula Br2Fe
Molecular Weight (g/mol) 215.65
MDL Number MFCD00016081
InChI Key GYCHYNMREWYSKH-UHFFFAOYSA-L
Synonym iron dibromide, unii-ea3x054rbz, iron ii bromide, ea3x054rbz, iron 2+ dibromide, iron ion 2+ dibromide, iron ii bromide, anhydrous 5g, iron ii bromide, ultra dry metals basis, ferrous bromide, anhydrous
PubChem CID 82240
IUPAC Name iron(2+);dibromide
SMILES [Fe++].[Br-].[Br-]

Specifications

Melting Point 684°C
Quantity 5 g
UN Number UN3260
Formula Weight 215.67
Percent Purity 99.995%
Packaging Ampouled under argon
Specific Gravity 4.636
Chemical Name or Material Iron(II) bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.