missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IRF5 (Human) Recombinant Protein
Human IRF5 full-length ORF ( AAH04139.1, 1 a.a. - 498 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003663-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MNQSIPVAPTPPRRVRLKPWLVAQVNSCQYPGLQWVNGEKKLFCIPWRHATRHGPSQDGDNTIFKAWAKETGKYTEGVDEADPAKWKANLRCALNKSRDFRLIYDGPRDMPPQPYKIYEVCSNGPAPTDSQPPEDYSFGAGEEEEEEEELQRMLPSLSLTEDVKWPPTLQPPTLRPPTLQPPTLQPPVVLGPPAPDPSPLAPPPGNPAGFRELLSEVLEPGPLPASLPPAGEQLLPDLLISPHMLPLTDLEIKFQYRGRPPRALTISNPHGCRLFYSQLEATQEQVELFGPISLEQVRFPSPEDIPSDKQRFYTNQLLDVLDRGLILQLQGQDLYAIRLCQCKVFWSGPCASAHDSCPNPIQREVKTKLFSLEHFLNELILFQKGQTNTPPPFEIFFCFGEEWPDRKPREKKLITVQVVPVAARLLLEMFSGELSWSADSIRLQISNPDLKDRMVEQFKELHHIWQSQQRLQPVAQAPPGAGLGVGQGPWPMHPAGMQ
Specifications
AAH04139.1 | |
interferon regulatory factor 5 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IRF5 | |
GST |
3663 | |
Wheat germ expression system | |
10 μg | |
SLEB10 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |