Learn More
Tocris Bioscience™ Irbesartan
Potent AT1 antagonist
Brand: Tocris Bioscience™ 5798/50
This item is not returnable.
View return policy
Description
Irbesartan is a potent angiotensin II type 1 receptor (AT1) antagonist (IC50 = 1.3 nM). Displays antihypertensive activity. Orally bioavailable.Specifications
| 2-Butyl-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1,3-diazaspiro[4.4]non-1-en-4-one | |
| 138402-11-6 | |
| 50 mg |